di-Kaliumtetraborat Tetrahydrat
Lieferant: Honeywell Chemicals
|
Gefahr
|
11620.9025
HONE11620.9025EA
0
EUR
HONE11620.9025
di-Kaliumtetraborat Tetrahydrat
di-Kaliumtetraborat Tetrahydrat
Formel:
B₄K₂O₇·4H₂O Dichte: 1,95 g/cm³ (20 °C) |
MDL:
MFCD00149924 CAS-Nummer: 12045-78-2 EINECS: 215-575-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...