Calciumhydroxid 95-100,5% (durch komplexometrische Analyse), puriss. FCC, Ph. Eur., USP, reinst, Fluka™

Lieferant: Honeywell Chemicals
Gefahr

12038.9025
HONE12038.9025EA 0 EUR
HONE12038.9025
Calciumhydroxid 95-100,5% (durch komplexometrische Analyse), puriss. FCC, Ph. Eur., USP, reinst, Fluka™
Calciumhydroxid
Formel: Ca(OH)₂
Schmelzpunkt: 550 °C
Dichte: 2,24 g/cm³
Lagertemperatur: Raumtemperatur
MDL: MFCD00010901
CAS-Nummer: 1305-62-0
EINECS: 215-137-3
ADR: 8,III
Merck Index: 13,01675

Bestellen

Spezifikation Testergebnisse

Assay (complexometric) 95.0 - 100.5 %
Assay (acidimetric) 95.0 - 100.5 %
Insoluble in hydrochloric acid Max. 0.2 %
Arsenic (As) Max. 3 ppm
Barium (Ba) Max. 300 ppm
Cadmium (Cd) Max. 1 ppm
Copper (Cu) Max. 0.001 %
Iron (Fe) Max. 0.1 %
Lead (Pb) Max. 1 ppm
Zinc (Zn) Max. 0.001 %
Heavy metals (as Pb) Max. 20 ppm
Mg and alkali salts (SO₄) Max. 2.5 %
Carbonate (CO₃) complying
Carbonate (as CaCO₃) Max. 5.0 %
Chloride (Cl) Max. 50 ppm
Fluoride (F) Max. 50 ppm
Sulfate (SO₄) Max. 0.05 %
Residual solvents complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR