Calciumchlorid Hexahydrat 97-103%, erfüllt analytische Spezifikationen von Ph. Eur., E509 (max. 0,0001% Al), Fluka™

Lieferant: Honeywell Chemicals
Achtung

12074.9025
HONE12074.9025EA 0 EUR
HONE12074.9025
Calciumchlorid Hexahydrat 97-103%, erfüllt analytische Spezifikationen von Ph. Eur., E509 (max. 0,0001% Al), Fluka™
Calciumchlorid Hexahydrat

  • Contains ≤0.0001% Al.

Formel: CaCl₂·6H₂O
Molecular Weight: 219,08 g/mol
Siedepunkt: 1,900 °C (1.013 hPa)
Schmelzpunkt: 29 °C
Dichte: 1,680 g/cm3 (20 °C)
Lagertemperatur: Gefrierschrank
MDL: MFCD00149616
CAS-Nummer: 7774-34-7
Merck Index: 13,01660

Bestellen

Spezifikation Testergebnisse

Transition Temp solidification point 28-30 °C
Identity complying
Assay 97.0 - 103.0 %
Solidification range 28 - 30 °C
Acid. or alk. react. impurities complying
Aluminium (Al) Max. 1 ppm
Arsenic (As) Max. 1 ppm
Barium (Ba) complying
Iron (Fe) Max. 5 ppm
Mercury (Hg) Max. 1 ppm
Lead (Pb) Max. 5 ppm
Heavy metals (as Pb) Max. 10 ppm
Magnesium and alkali metals Max. 0.3 %
Fluoride (F) Max. 40 ppm
Sulfate (SO₄) Max. 100 ppm
Residual solvents complying
Appearance of the solution complying

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR