Chrom(III)chlorid Hexahydrat
Lieferant: Honeywell Chemicals
|
Achtung
|
Synonyme:
Chromtrichlorid Hexahydrat
12228.9025
HONE12228.9025EA
0
EUR
HONE12228.9025
Chrom(III)chlorid Hexahydrat
Chrom(III)chlorid Hexahydrat
Formel:
CrCl₃·6H₂O Molecular Weight: 266.45 g/mol Schmelzpunkt: 85 Dichte: 1.76 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149660 CAS-Nummer: 10060-12-5 EINECS: 233-038-3 UN: 3260 ADR: 8 III Merck Index: 13,02242 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...