Chrom(III)nitrat Nonahydrat
Lieferant: Honeywell Chemicals
|
Achtung
|
Synonyme:
Chromtrinitrat Nonahydrat
12232.9025
HONE12232.9025EA
0
EUR
HONE12232.9025
Chrom(III)nitrat Nonahydrat
Chrom(III)nitrat Nonahydrat
Formel:
CrN₃O₉·9H₂O Schmelzpunkt: 60 °C Dichte: 1,807 g/cm³ (20 °C) |
MDL:
MFCD00149659 CAS-Nummer: 7789-02-8 EINECS: 236-921-1 UN: 2720 ADR: 5.1,III Merck Index: 13,02246 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...