Chrom(III)nitrat 65% in wässriger Lösung

Lieferant: Honeywell Chemicals

12278.9075
HONE12278.9075EA 0 EUR
HONE12278.9075
Chrom(III)nitrat 65% in wässriger Lösung
Chrom(III)nitrat
Formel: CrN₃O₉
CAS-Nummer: 13548-38-4
EINECS: 236-921-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR