Magnesiumhydroxidcarbonat leicht (∼40,0 - 45,0% MgO), Pulver, puriss. Ph. Eur., BP, E504, Fluka™

Lieferant: Honeywell Chemicals

Synonyme: Magnesiumcarbonat basisch, Magnesiumcarbonathydroxid, Magnesiumcarbonat basisch hydratisiert

13117.9020
HONE13117.9020EA 0 EUR
HONE13117.9020
Magnesiumhydroxidcarbonat leicht (∼40,0 - 45,0% MgO), Pulver, puriss. Ph. Eur., BP, E504, Fluka™
Magnesiumhydroxidcarbonat
Formel: MgCO₃ (linear)
Molecular Weight: 84,31 g/mol
Dichte: 2,16 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149783
CAS-Nummer: 12125-28-9
EINECS: 235-192-7

Bestellen

Spezifikation Testergebnisse

Assay of Mg (as MgO) 40.0 - 45.0 %
In water soluble matter Max. 1.0 %
Insoluble in acetic acid Max. 0.05 %
Arsenic (As) Max. 1 ppm
Calcium (Ca) Max. 0.4 %
Copper (Cu) Max. 0.001 %
Iron (Fe) Max. 200 ppm
Mercury (Hg) Max. 1 ppm
Lead (Pb) Max. 2 ppm
Zinc (Zn) Max. 0.001 %
Heavy metals (as Pb) Max. 10 ppm
Chloride (Cl) Max. 300 ppm
Sulfate (SO₄) Max. 0.2 %
Residual solvents complying
Appearance of the solution complying

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR