Magnesiumsulfat Heptahydrat 99,5-100,5 Dried, meetSpec_BP,Ph.Eur.,USP,FCC, Fluka™

Lieferant: Honeywell Chemicals

Synonyme: Bittersalz

13142-5KG 13142-25KG 13142.9025 13142-1KG 13142-4X5KG
13142-5KGEA 108.91 EUR
13142-5KG 13142-25KG HONE13142.9025 13142-1KG 13142-4X5KG
Magnesiumsulfat Heptahydrat 99,5-100,5 Dried, meetSpec_BP,Ph.Eur.,USP,FCC, Fluka™
Magnesiumsulfat Heptahydrat
Formel: MgSO₄·7H₂O
Molecular Weight: 246.48 g/mol
Schmelzpunkt: 1.124 °C (Zers.)
Dichte: 1.68 g/cm³ at 20 °C
Lagertemperatur: Room Temperature
MDL: MFCD00149785
CAS-Nummer: 10034-99-8
EINECS: 231-298-2
Merck Index: 13,05715

Bestellen

Spezifikation Testergebnisse

Assay (calculated to the dried substance) 99.5 - 100.5 %
Loss on drying (450°C) 48.0 - 52.0 %
pH (5 %, 25 °C) 5.0 - 9.2
Free acid (as H₂SO₄) Max. 0.01 %
Free alkali (as MgO) Max. 0.004 %
Acidity or alkalinity complying
Arsenic (As) Max. 2 ppm
Iron (Fe) Max. 20 ppm
Lead (Pb) Max. 4 ppm
Selenium (Se) Max. 30 ppm
Heavy metals (as Pb) Max. 10 ppm
Chloride (Cl) Max. 100 ppm
Residual solvents complying
Appearance of the solution complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR