Mangan(II)carbonat Hydrat

Lieferant: Honeywell Chemicals

Synonyme: Mangancarbonat Hydrat

13215.9025
HONE13215.9025EA 0 EUR
HONE13215.9025
Mangan(II)carbonat Hydrat
Mangan(II)carbonat Hydrat
Formel: CMnO₃·H₂O
Molecular Weight: 132,96 g/mol
CAS-Nummer: 34156-69-9
EINECS: 209-942-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR