Mangan(II)chlorid Monohydrat, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
M-Clarity™ Qualitätsstufe = MQ200
13220-2.5KG-R
13220.9030
SIAL13220-2.5KG-REA
180
EUR
SIAL13220-2.5KG-R
HONE13220.9030
Mangan(II)chlorid Monohydrat, Sigma-Aldrich®
Mangan(II)chlorid Monohydrat
Formel:
Cl₂Mn·1H₂O Molecular Weight: 143.86 g/mol Lagertemperatur: Raumtemperatur |
CAS-Nummer:
64333-01-3 EINECS: 231-869-6 UN: 3077 ADR: 9 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...