Mangan(IV)oxid
Lieferant: Honeywell Chemicals
|
Gefahr
|
Synonyme:
Mangandioxid, Pyrolusit, Braunstein
13242.9050
HONE13242.9050EA
0
EUR
HONE13242.9050
Mangan(IV)oxid
Mangan(IV)oxid
Formel:
MnO₂ Molecular Weight: 86.94 g/mol Schmelzpunkt: 535 Dichte: 5.03 g/cm³ (20 °C) |
MDL:
MFCD00003463 CAS-Nummer: 1313-13-9 EINECS: 215-202-6 Merck Index: 14,05730 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...