Mangan(II)acetat Tetrahydrat

Lieferant: Honeywell Chemicals

13286.9025
HONE13286.9025EA 643 EUR
HONE13286.9025
Mangan(II)acetat Tetrahydrat
Mangan(II)acetat Tetrahydrat
Formel: (H₃CCOO)₂Mn·4H₂O
Schmelzpunkt: 300 °C
Dichte: 1,58 g/cm³ (20 °C)
MDL: MFCD00062552
CAS-Nummer: 6156-78-1
EINECS: 211-334-3
Merck Index: 13,05746

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR