Natriumstannat SnO₂ 42-45%

Lieferant: Honeywell Chemicals
Achtung

14505.9050
HONE14505.9050EA 0 EUR
HONE14505.9050
Natriumstannat SnO₂ 42-45%
Natriumstannat
Formel: Na₂O₃Sn
Dichte: 4,68 g/cm³ (25 °C)
MDL: MFCD00014241
CAS-Nummer: 12058-66-1
EINECS: 235-030-5
Merck Index: 13,08752

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR