Zinn(II)sulfat

Lieferant: Honeywell Chemicals

Synonyme: Zinnsulfat

14531.9050
HONE14531.9050EA 0 EUR
HONE14531.9050
Zinn(II)sulfat
Zinn(II)sulfat
Formel: O₄SSn
Molecular Weight: 214,77 g/mol
Dichte: 4,15 g/cm³ (20 °C)
MDL: MFCD00011246
CAS-Nummer: 7488-55-3
EINECS: 231-302-2
Merck Index: 12,08947

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR