Tetrabutylammoniumfluorid

Lieferant: Honeywell Chemicals

16279.9015
HONE16279.9015EA 0 EUR
HONE16279.9015
Tetrabutylammoniumfluorid
Tetrabutylammoniumfluorid
Formel: C₁₆H₃₆FN
Dichte: 0,903 g/cm³ (20 °C)
MDL: MFCD00011747
CAS-Nummer: 429-41-4
EINECS: 207-057-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR