Tetramethylammoniumhydroxid 25% aqueous solution

Lieferant: Honeywell Chemicals
Gefahr

17032.5000
HONE17032.5000EA 0 EUR
HONE17032.5000
Tetramethylammoniumhydroxid 25% aqueous solution
Tetramethylammonium hydroxide
Formel: C₄H₁₃NO
Molecular Weight: 91.15 g/mol
Dichte: 1.02 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008280
CAS-Nummer: 75-59-2
EINECS: 200-882-9
UN: 1835
ADR: 8 II
Merck Index: 14,09224

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR