Ätzmischung AF 87,5-12,5 87.5-12.5
Lieferant: Honeywell Chemicals
|
Gefahr
|
Synonyme:
Ammonium fluoride - hydrofluoric acid mixture (87.5:12.5)
17621.5000
HONE17621.5000EA
0
EUR
HONE17621.5000
Ätzmischung AF 87,5-12,5 87.5-12.5
Ätzmischung AF 87,5-12,5
UN:
2922 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...