Natriumacetat Trihydrat 99,0-101,0% (Trockenmasse), Puriss., erfüllt analytische Spezifikationen von BP, FCC, Ph. Eur., USP, E262 (max. 0,00002% Al), Fluka™

Lieferant: Honeywell Chemicals

25022-1KG 25022-5KG 25022-25KG 25022.9025 25022-2.5KG 25022-6X1KG
25022-1KGEA 58.62 EUR
25022-1KG 25022-5KG 25022-25KG HONE25022.9025 25022-2.5KG 25022-6X1KG
Natriumacetat Trihydrat 99,0-101,0% (Trockenmasse), Puriss., erfüllt analytische Spezifikationen von BP, FCC, Ph. Eur., USP, E262 (max. 0,00002% Al), Fluka™
Natriumacetat Trihydrat
Formel: H₃CCOONa·3H₂O
Schmelzpunkt: 58 °C
Dichte: 1,45 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00071557
CAS-Nummer: 6131-90-4
EINECS: 204-823-8
Merck Index: 13,08642
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=313c1d45673fab192565649f8cbb7ddff555de39&vid=1ad37a966605626c95117735c83b16eaea381fbc&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay (calculated to the dried substance) 99.0 - 101.0 %
Loss on drying (130°C) 39.0 - 40.5 %
pH (5 %, 25 °C) 7.5 - 9.0
Water insoluble matter Max. 0.05 %
Free alkali (as Na₂CO₃) Max. 0.05 %
Aluminium (Al) Max. 0.2 ppm
Arsenic (As) Max. 2 ppm
Calcium (Ca) Max. 20 ppm
Calcium, Magnesium complying
Copper (Cu) Max. 0.001 %
Iron (Fe) Max. 10 ppm
Mercury (Hg) Max. 1 ppm
Potassium (K) complying
Magnesium (Mg) Max. 20 ppm
Lead (Pb) Max. 2 ppm
Zinc (Zn) Max. 0.001 %
Heavy metals (as Pb) Max. 10 ppm
Chloride (Cl) Max. 50 ppm
Sulfate (SO₄) Max. 50 ppm
KMnO₄ reducing matter (as HCOOH) Max. 0.1 %
KMnO₄ reducing matter (as O) Max. 0.002 %
Formic acid, formiates other impurities (HCOOH) Max. 1000 ppm
Appearance of the solution complying
Reducing impurities complying
Residual solvents complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR