(4-Bromphenyl)essigsäure
Lieferant: Honeywell Chemicals
65166.9025
HONE65166.9025EA
0
EUR
HONE65166.9025
(4-Bromphenyl)essigsäure
(4-Bromphenyl)essigsäure
Formel:
(C₆H₄Br)CH₂COOH Siedepunkt: 326 °C (1013 hPa) Schmelzpunkt: 94…98 °C |
MDL:
MFCD00004342 CAS-Nummer: 1878-68-8 EINECS: 217-523-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...