Shoe Covers, PP Spunlaid Non Woven + PE Sole Plastic-Coated Sole
Lieferant: Medicom
10.012LONG
HOPE10.012LONGEA 94.6 EUR
HOPE10.012LONG
Shoe Covers, PP Spunlaid Non Woven + PE Sole Plastic-Coated Sole
Einweg-Kleidung
Overshoes PP spunlaid non woven with PE sole Plastic-coated sole.
  • Overshoe with non-slip sole
  • They adapt to different morphologies and all types of shoes
  • Latex-free elastic
Standards: ISO 9001 Manufacturing site.