Tape, high temperature
Lieferant: HORST
100102
HORS100102EA 51.5 EUR
HORS100102
Tape, high temperature
Klebebänder
This high quality adhesive tape is made from glass fabric with silicone adhesive and is suitable for the mounting of heating cables up to 200 °C.
  • Corresponds to class H (180 °C)
Length: 50,0 m
Colour: White