Degassing Channel tubing: systec af (0.045 id) Degassing Channel Pressure rating: 70 Psi (testing pressure) wetted Materials: systec af, PPs, glass-filled PtfE, stainless steel Liquid Connection: 1/4-28 unf threaded flat-bottom port Size (L x H 1 * 1 ST
Lieferant: COLLECTION PREFIX HPLC
Degassing Channel tubing: systec af (0.045 id) Degassing Channel Pressure rating: 70 Psi (testing pressure) wetted Materials: systec af, PPs, glass-filled PtfE, stainless steel Liquid Connection: 1/4-28 unf threaded flat-bottom port Size (L x H 1 * 1 ST