Column Oven CO20 - Easy to use single Column heater with temperature range from room temperature to 90,0C - Holds 1-30 cm long by 1/4 or 3/8 diameter column - Stable to +/- 0,1C - Accurate to +/- 0,2C - Chamber size 381 mm long x 22,9 mm wide x 1 * 1 ST
Lieferant: COLLECTION PREFIX HPLC
Column Oven CO20 - Easy to use single Column heater with temperature range from room temperature to 90,0C - Holds 1-30 cm long by 1/4 or 3/8 diameter column - Stable to +/- 0,1C - Accurate to +/- 0,2C - Chamber size 381 mm long x 22,9 mm wide x 1 * 1 ST