Thermo Scientific™ Teleshake 1536-8 Magnetic Microtiter Plate Shaker
Lieferant: THERMO H + P LABORTECHNIK
50101683
HPLT50101683EA 5240 EUR
HPLT50101683
Thermo Scientific™ Teleshake 1536-8 Magnetic Microtiter Plate Shaker
Schüttler Mikrotiterplatten-Schüttler
Perform powerful resonance shaking up to 8500 min⁻¹ with Thermo Scientific™ Teleshake 1536-8 Magnetic Microtiter Plate Shakers, which feature a wear-free magnetic drive. They are remote controlled for integrated shaking applications, and are suited for 1536- and 384-well microtiter plates.
  • Acceleration up to 40×g in the resonance range with a minimum of moved masses which translates into little transference of vibrations onto the respective installation areas
  • Easy integration in robotic systems and platform due to small footprint
  • Simultaneous control of up to 14 shakers via RS232 interface
  • Rapid, ultra-quiet shaking at up to 40×g in the resonance range
  • Adjustable amplitude from 0,1 to 1 mm
  • Available with 6 or 8-position fixing angles Integrated threads and drilled holes for assembling on robot workbenches
  • Electromechanical mechanism locks down microplates safely