Sodium borate-Lösung 5 g/dl in deionisiertem Wasser
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Gefahr
|
M-Clarity™ Qualitätsstufe = MQ500
HT1002-100ML
HT1002-100MLEA
108
EUR
HT1002-100ML
Sodium borate-Lösung 5 g/dl in deionisiertem Wasser
Sodium borate solutions
Kit component in the silver stain kit.
Spezifikation Testergebnisse
Appearance (Colour) | Colorless |
Appearance (Form) | Solution |
Appearance (Turbidity) Essentially free of particles/precipitate |
Clear |
Performance When using Sigma-Aldrich Procedure No. HT100, fungi organisms stain purplish-brown to black |
Conforms |
pH | 8.9 - 9.6 |
Expiration Date Period | 24 months |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...