Gold chloride solution_SC 200 mg/dl water_deionized
Lieferant: SIGMA ALDRICH MICROSCOPY
MQ500
HT1004-100ML
HT1004-100MLEA
122
EUR
HT1004-100ML
Gold chloride solution_SC 200 mg/dl water_deionized
Gold chloride
Used in procedure HT100 for the toning of sections stained with the silver methanamine procedure.
- Gold chloride, 0.2% in aqueous solution
|
Spezifikation Testergebnisse
Appearance (Colour) | Yellow |
Appearance (Form) | Solution |
Appearance (Turbidity) Essentially free of particles/precipitate |
Clear |
Performance | Conforms |