Natriumthiosulfat 20 g/L in wässriger Lösung, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
Synonyme:
di-Natriumthiosulfat, Antichlor, Fixiersalz, Natriumhyposulfit
HT1005-100ML
HT1005-100MLEA
106
EUR
HT1005-100ML
Natriumthiosulfat 20 g/L in wässriger Lösung, Sigma-Aldrich®
Natriumthiosulfat
Formel:
Na₂S₂O₃ Molecular Weight: 158.11 g/mol Siedepunkt: 100 Schmelzpunkt: 48 Dichte: 1010 g/cm³ Lagertemperatur: Kühlschrank |
MDL:
MFCD00003499 CAS-Nummer: 7772-98-7 EINECS: 231-867-5 Merck Index: 12,08844 |
|
Spezifikation Testergebnisse
Appearance (Colour) | Colourless |
Appearance (Form) | Solution |
Appearance (Turbidity), Essentially free of particles/precipitate | Clear |
Performance | Conforms |
Expiration Date Period | 24 months |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...