Silver Stain (Modified GMS) Kit, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr

Synonyme: Modified Grocott Methanamine Silver (GMS) Fungus Stain, Periodic Acid Methanamine Silver Technique

MQ500

HT100A-1KT
HT100A-1KTEA 518 EUR
HT100A-1KT
Silver Stain (Modified GMS) Kit, Sigma-Aldrich®
Silver Stain (Modified GMS) Kit

Silver stain is an aid in distinguishing fungi, basement membrane and opportunistic organisms such as Pneumocystis carinii in tissue sections or smears.


  • Suitable for haematology
  • Suitable for histology

Lagertemperatur: Kühlschrank
UN: 3316
IVDLogo

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR