Hydrochinon
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Gefahr
|
M-Clarity™ Qualitätsstufe = MQ500
HT1014-10TAB
HT1014-10TABEA
158
EUR
HT1014-10TAB
Hydrochinon
Hydrochinon
Intended for use in the Silver Stain (Steiner-Steiner) kit. Hydroquinone, 0.5 g each, with excipients.
Formel:
C₆H₄(OH)₂ Flammpunkt: 165 °C (geschlossener Tiegel) Lagertemperatur: Kühlschrank |
MDL:
MFCD00002339 CAS-Nummer: 123-31-9 |
Spezifikation Testergebnisse
Appearance (Colour) | White to Off White |
Appearance (Form) Intact |
Round Tablet |
Performance | Conforms |
Expiration Date Period | 24 months |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...