Trichrome stain solution LG, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
Synonyme: Gomori Trichrome
HT10316-500ML
HT10316-500MLEA 502 EUR
HT10316-500ML
Trichrome stain solution LG, Sigma-Aldrich®
Trichrome stain
Intended for use in the study of connective tissue, muscle and collagen fibers. Stains collagen green.
IVDLogo