Trichrome stain solution AB, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
Synonyme:
Gomori Trichrome
HT10516-500ML
HT10516-500MLEA
502
EUR
HT10516-500ML
Trichrome stain solution AB, Sigma-Aldrich®
Trichrome stain
Intended for use in the study of connective tissue, muscle and collagen fibers. Stains collagen blue and muscle red.