Eisenhämatoxylin, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Gefahr
|
Synonyme:
Weigert's solution, Weigert's stain
M-Clarity™ Qualitätsstufe = MQ500
HT1079-1SET
HT1079-1SETEA
235
EUR
HT1079-1SET
Eisenhämatoxylin, Sigma-Aldrich®
Eisenhämatoxylin
Recommended in lengthy or acidic stain procedures. Resists decolorisation better than aluminum based hematoxylin formulations. Weigert's iron hematoxylin solution set consists of HT107 Part A and HT109 Part B.
Spezifikation Testergebnisse
Certificate of Analysis | Accept |
The individual components of this kit conforms to the Sigma-Aldrich quality testing and performance specifications as outlined in the package insert. | |
Expiration Date Period | Earliest component expiration date |