Eisenhämatoxylin-Lösung (Weigert) Part B, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

Synonyme: Weigert's solution, Weigert's stain

HT109-500ML
HT109-500MLEA 126 EUR
HT109-500ML
Eisenhämatoxylin-Lösung (Weigert) Part B, Sigma-Aldrich®
Eisenhämatoxylin

To be used in conjunction with HT107. Recommended in lengthy or acidic stain procedures. Resists decolorization better than aluminum based hematoxylin formulations.

Lagertemperatur: Raumtemperatur
MDL: MFCD00284879
ADR: 8,III
IVDLogo

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR