Eosin Y 0,5 % (w/v) alkoholische Lösung in acidified ethanol, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr


M-Clarity™ Qualitätsstufe = MQ500

HT110132-1L
HT110132-1LEA 113 EUR
HT110132-1L
Eosin Y 0,5 % (w/v) alkoholische Lösung in acidified ethanol, Sigma-Aldrich®
Eosin Y

General purpose cytoplasmic counterstain. Used with hematoxylin and eosin staining.


Standards: Certified Eosin Y, 0.5% (w/v) in acidified 90% ethanol.

Formel: C₂₀H₈Br₄O₅
Flammpunkt: 13 °C (geschlossener Tiegel)
Lagertemperatur: Room Temperature
MDL: MFCD00036189
CAS-Nummer: 15086-94-9

Bestellen

Spezifikation Testergebnisse

Appearance (Colour)
Bright Pink to Pinkish-Orange
Conforms
Appearance (Form) Solution
Appearance (Turbidity) Clear
Performance Conforms
When using Sigma-Aldrich Procedure No. HT110-1, tissue cytoplasm stains pinkish-orange to pink with 3 tone effect noted. Nuclei stain blue to black due to Hematoxylin.
Expiration Date Period 18 months


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR