Eosin Y-Lösung Aqueous
Lieferant: SIGMA ALDRICH MICROSCOPY
Synonyme:
2-(3,6-Dihydroxy-2,4,5,7-tetrabromxanthen-9-yl)-benzoesäure, Tetrabromfluorescein, 2,4,5,7-Tetrabromfluorescein, 2′,4′,5′,7′-Tetrabrom-3′,6′-dihydroxyspiro [2-benzofuran-3,9′-xanthen]-1-on
M-Clarity™ Qualitätsstufe = MQ500
HT1102128-4L
HT1102128-4LEA
320
EUR
HT1102128-4L
Eosin Y-Lösung Aqueous
Eosin Y
General purpose cytoplasmic counterstain. Used with hematoxylin and eosin staining.
Standards: Certified Eosin Y, 0.5% (w/v) in water. Not acidified.
Formel:
C₂₀H₈Br₄O₅ Molecular Weight: 647,89 g/mol Lagertemperatur: Room Temperature |
MDL:
MFCD00036189 CAS-Nummer: 15086-94-9 |
Spezifikation Testergebnisse
Appearance (Colour) Greenish- Pink-Orange |
Conforms |
Appearance (Form) | Solution |
Appearance (Turbidity) Essentially free of precipitate |
Clear |
Performance | Conforms |
Cytoplasm stains pinkish-orange to pink with a 3 tone effect | |
Expiration Date Period | 18 months |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...