Eosin Y solution_SC Aqueous, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyme: 2-(3,6-Dihydroxy-2,4,5,7-tetrabromxanthen-9-yl)-benzoesäure, Tetrabromfluorescein, 2,4,5,7-Tetrabromfluorescein, 2′,4′,5′,7′-Tetrabrom-3′,6′-dihydroxyspiro [2-benzofuran-3,9′-xanthen]-1-on

MQ500

HT110232-1L
HT110232-1LEA 106 EUR
HT110232-1L
Eosin Y solution_SC Aqueous, Sigma-Aldrich®
Eosin Y

General purpose cytoplasmic counterstain. Used with hematoxylin and eosin staining.


Standards: Certified Eosin Y, 0.5% (w/v) in water. Not acidified.

Formel: C₂₀H₈Br₄O₅
Molecular Weight: 647.89 g/mol
Schmelzpunkt: 295
Lagertemperatur: Room Temperature
MDL: MFCD00036189
CAS-Nummer: 15086-94-9

Bestellen

Spezifikation Testergebnisse

Appearance (Colour)
Greenish- Pink-Orange
Conforms
Appearance (Form) Solution
Appearance (Turbidity) Clear
Essentially free of precipitate
Performance Conforms
Cytoplasm stains pinkish-orange to pink with a 3 tone effect
Expiration Date Period 18 months


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR