Eosin Y-Lösung, alkoholisch , with Phloxin

Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr

Synonyme: 2-(3,6-Dihydroxy-2,4,5,7-tetrabromxanthen-9-yl)-benzoesäure, Tetrabromfluorescein, 2,4,5,7-Tetrabromfluorescein, 2′,4′,5′,7′-Tetrabrom-3′,6′-dihydroxyspiro [2-benzofuran-3,9′-xanthen]-1-on

M-Clarity™ Qualitätsstufe = MQ500

HT1103128-4L
HT1103128-4LEA 330 EUR
HT1103128-4L
Eosin Y-Lösung, alkoholisch , with Phloxin
Eosin Y

General purpose cytoplasmic counterstain. Used with hematoxylin and eosin staining.


  • Certified Eosin Y, 0.1% (w/v) and certified phloxine B, 0.1% (w/v) in acidified ethanol

Formel: C₂₀H₈Br₄O₅
Molecular Weight: 647,89 g/mol
Flammpunkt: 13 °C (geschlossener Tiegel)
Lagertemperatur: Room Temperature
MDL: MFCD00036189
CAS-Nummer: 15086-94-9

Bestellen

Spezifikation Testergebnisse

Appearance (Colour)
Bright pink to pinkish-orange
Conforms
Appearance (Form) Solution
Appearance (Turbidity) Clear
Essentially free of particles
Performance Conforms
When using with Sigma-Aldrich Procedure No. HT110-3, Cytoplasm stains pinkish-orange to pink with 3 tone effect noted.
Expiration Date Period 24 months

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR