Papanicolaou's Lösung (OG 6), Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr


MQ500

HT40116-500ML HT40132-1L
HT40116-500MLEA 52 EUR
HT40116-500ML HT40132-1L
Papanicolaou's Lösung (OG 6), Sigma-Aldrich®
Papanicolaou's Lösung (OG 6)

Orange G (certified), 0.3% and phosphotungstic acid, 0.015%, in denatured alcohol


Papanicolaou stain imparts a characteristic range of coloration to exfoliative cells of vaginal, cervical, prostatic and other body secretions, allowing critical examination of nuclei and cytoplasmic components.

Lagertemperatur: Raumtemperatur
MDL: MFCD00147833
UN: 1170
ADR: 3 II
IVDLogo

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR