Papanicolaou's solution modified EA, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Gefahr
|
HT40232-1L
HT40232-1LEA
86.7
EUR
HT40232-1L
Papanicolaou's solution modified EA, Sigma-Aldrich®
Papanicolaou's solution modified
Papanicolaou stain imparts a characteristic range of coloration to exfoliative cells of vaginal, cervical, prostatic and other body secretions, allowing critical examination of nuclei and cytoplasmic components.
Lagertemperatur:
Raumtemperatur |
ADR:
3,II |
![]() |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...