Papanicolaou's solution EA 65, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

Synonyme: EA 36

HT40432-1L
HT40432-1LEA 136 EUR
HT40432-1L
Papanicolaou's solution EA 65, Sigma-Aldrich®
Papanicolaou's solution EA 65

Papanicolaou stain imparts a characteristic range of coloration to exfoliative cells of vaginal, cervical, prostatic and other body secretions, allowing critical examination of nuclei and cytoplasmic components.

Lagertemperatur: Raumtemperatur
ADR: 3,II
IVDLogo

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR