Formalinlösung 10%, gepuffert (neutral) (pH 7.0 ± 0.2), Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Achtung
|
Synonyme:
Formaldehyd, Methanal, Formol, Methylaldehyd
M-Clarity™ Qualitätsstufe = MQ200
HT501128-4L
HT501128-4LEA
87
EUR
HT501128-4L
Formalinlösung 10%, gepuffert (neutral) (pH 7.0 ± 0.2), Sigma-Aldrich®
Formalinlösung
(= 10% Formaline solution)
Formel:
CH₂O Molecular Weight: 30,03 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003274 CAS-Nummer: 50-00-0 EINECS: 200-001-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...