Formalinlösung 10% Fixativ für die Histologie, neutral gepuffert, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY


M-Clarity™ Qualitätsstufe = MQ200

HT5012-1CS
HT5012-1CSEA 191 EUR
HT5012-1CS
Formalinlösung 10% Fixativ für die Histologie, neutral gepuffert, Sigma-Aldrich®
Formalinlösung

Formalin solution (10%, neutral buffered) is a widely used fixative for samples processed for light microscopy. It requires a relatively short period of fixation. It is suitable for the long-term storage of tissue. Once properly fixed, the tissue should be able to withstand the subsequent stages of tissue processing or staining.


  • Formalin solution (neutral buffered, 10%) has been popularly used as a general-purpose histological fixative to fix tissue samples
  • Popular general purpose histological fixative


Verpackung: Case containers are polypropylene with white leak-proof lids and are partially prefilled with 10% NBF.

Formel: CH₂O
Dichte: 1,080 g/cm3
Lagertemperatur: Room Temperature
MDL: MFCD00003274
CAS-Nummer: 50-00-0

Bestellen

Spezifikation Testergebnisse

Appearance (Colour) Colorless
Appearance (Turbidity) Clear
Conforms Performance
Fixation test within acceptable limits
pH 6.90 - 7.10


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR