Formalinlösung-Lösung 10% (synthetisch), neutral gepuffert, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
Synonyme:
Formaldehyd, Methanal, Formol, Methylaldehyd
HT501320-9.5L
HT501320-9.5LEA
147
EUR
HT501320-9.5L
Formalinlösung-Lösung 10% (synthetisch), neutral gepuffert, Sigma-Aldrich®
Formalinlösung
Formalin solution, neutral buffered, 10% is the most commonly used fixative for light microscopy. It requires a relatively short period of fixation. It is suitable for the long-term storage of tissue. Once properly fixed, the tissue should be able to withstand the subsequent stages of tissue processing or staining.
- Formalin solution, neutral buffered, 10% has been used as a popular general purpose histological fixative to fix tissues
- Popular general purpose histological fixative
Verpackung: Item is packaged in a 10 L cubitainer consisting of a plastic bladder inside of a cardboard box with an attached spigot.
Formel:
CH₂O Molecular Weight: 30,03 g/mol Dichte: 1,080 g/cm3 Lagertemperatur: Room Temperature |
MDL:
MFCD00003274 CAS-Nummer: 50-00-0 |
Spezifikation Testergebnisse
Appearance (Colour) | Colorless |
Appearance (Turbidity) | Clear |
Performance | Pass |
Fixation test within acceptable limits | |
pH | 6.90 - 7.10 |
Expiration Date Period | 30 months |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...