Amyloid Stain, Congo Red Kit, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Gefahr
|
M-Clarity™ Qualitätsstufe = MQ500
Amyloid protein is detected in tissue with Congo red, a metachromatic stain. Staining is intensified by pretreatment of tissue with alkaline sodium chloride.
- Suitable for haematology
- Suitable for histology
Kit component only
Congo Red Solution (kit only) 500 mL
Sodium Chloride Solution, Alcoholic (kit only) 500 mL
Sodium Hydroxide Solution (kit only) 2 x 12 mL
Lagertemperatur:
Kühlschrank |
UN:
2924 |
![]() |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...