Methyl Green-Pyronin Y-Lösung, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
Synonyme:
MGP
HT70116-500ML
HT70116-500MLEA
440
EUR
HT70116-500ML
Methyl Green-Pyronin Y-Lösung, Sigma-Aldrich®
Methyl Green-Pyronin Y
Pyronin stains the cytoplasm of plasma cells and most nucleoli red.