Karbolfuchsin-Lösung, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr

Synonyme: 4-[(4-Amino-m-tolyl)(4-iminocyclohexa-2,5-dien-1-yliden)methyl]-o-toluidin Hydrochlorid, 4-[(4-Amino-m-tolyl)(4-iminocyclohexa-2,5-dien-1-yliden)methyl]-o-toluidiniumchlorid, 4-(Di(4-amino-3-methylphenyl)methylen)-2,5-cyclohexadien-1-yliden)ammoniumchlorid, 3,3'-Dimethyl-4,4',4''-triaminotrityliumchlorid

M-Clarity™ Qualitätsstufe = MQ500

HT80116-500ML
HT80116-500MLEA 343 EUR
HT80116-500ML
Karbolfuchsin-Lösung, Sigma-Aldrich®
Karbolfuchsin

Intended for use in the Acid Fast Stain, Procedure No. HT80 with Malachite Green counterstain Catalog No. HT80-2


  • Pararosaniline (certified), 0.85%; phenol, 2.5%; glycerol, 5%; DMSO, 5%, and ethyl alcohol, 5%, in deionized water

Formel: C₂₁H₂₂ClN₃
Molecular Weight: 351,88 g/mol
Lagertemperatur: Room Temperature
MDL: MFCD00013129
CAS-Nummer: 4197-24-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=510b48d87a0933ee2858d800b355527793898b42&vid=00afa7f8871ec798fd3912fd1c9b37e00466987a&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Colour)
Dark red-purple with metallic sheen
Conforms
Appearance (Form) Solution
Appearance (Turbidity) Clear
Wave Length 542 - 552 nm
Absorbance 0.300 - 0.700
Performance Conforms
Acid Fast Bacilli Red; Background green when performing with Sigma-Aldrich Procedure HT 80
Expiration Date Period
18 months
Complete

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR