Karbolfuchsin-Lösung, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Gefahr
|
HT8018-250ML
HT8018-250MLEA
193
EUR
HT8018-250ML
Karbolfuchsin-Lösung, Sigma-Aldrich®
Karbolfuchsin
Intended for use in the Acid Fast Stain, Procedure No. HT80 with Malachite Green counterstain Catalog No. HT80-2
Formel:
C₂₁H₂₂ClN₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013129 CAS-Nummer: 4197-24-4 |
|
![]() |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...