Kristallviolett-Lösung, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

Synonyme: Hexamethyl-p-rosaniliniumchlorid, Hexamethylen-p-rosanilinchlorid, blaues Pyoktanin, Basic Violet 3, Hexamethylviolett, 4-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]dimethylammoniumchlorid, Methylviolett 10B, Methylrosanilin, Methylrosanilium, 4,4',4''-Tri(dimethylamino)trityliumchlorid, CI 42555, Methylrosaniliumchlorid, Gentianaviolett

HT90132-1L
HT90132-1LEA 122 EUR
HT90132-1L
Kristallviolett-Lösung, Sigma-Aldrich®
Kristallviolett

Intended for use in the Gram Stain Procedure No. HT90.

Formel: C₂₅H₃₀ClN₃
Molecular Weight: 407,99 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00011750
CAS-Nummer: 548-62-9
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce585fad352839510d2a1a590f07fa766ee8e97d&vid=d52b57138a4d90e1a26aab603af051f7240eeb7e&prid=web&clid=SAPDAM IVDLogo

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR