Gram-color Färbekit für Folien, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Gefahr
|
HT90A-1KT
HT90A-1KTEA
209
EUR
HT90A-1KT
Gram-color Färbekit für Folien, Sigma-Aldrich®
Gram color staining kit
Films are stained for Gram-positive organisms with crystal violet, mordanted in iodine, decolorized and then stained for Gram-negative organisms with safranin O. For delineating Gram-positive and Gram-negative organisms in films.
Lagertemperatur:
Raumtemperatur |
ADR:
9,II |
![]() |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...