Gram-color Färbekit für Gewebe, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Gefahr
|
HT90T-1KT
HT90T-1KTEA
279
EUR
HT90T-1KT
Gram-color Färbekit für Gewebe, Sigma-Aldrich®
Gram color staining kit
Tissues are stained for Gram-positive organisms with crystal violet, mordanted in iodine, decolorized, stained for Gram-negative organisms with safranin O and then counterstained with tartrazine. For delineating Gram-positive and Gram-negative organisms
Lagertemperatur:
Raumtemperatur |
ADR:
9,III |
![]() |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...